• USA Home
  • Search Results

  • ethanethiol62137508111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ethanethiol

4 Products
Synonym:   Ethyl mercaptan
Image of Ethanethiol
CAS Number:
Empirical Formula (Hill Notation):
 C2H6S
Molecular Weight:
 62.13
 EC Index Number
 200-837-3
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Special Grade
  Titertest
Show More