• USA Home
  • Search Results

  • ethanol46076417511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ethanol

10 Products
Synonym:   Ethyl alcohol
Image of Ethanol
CAS Number:
Linear Formula:
 CH3CH2OH
Molecular Weight:
 46.07
 Beilstein/REAXYS Number:
 1718733
 EC Index Number:
 200-578-6
 MDL number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Additive
  Shelf Life
  ID clarifier
  Application(s)
  Color
  Physical Form
  Absorption
  Solubility
  Density
  Agency/Method
  pH Value
  UV-Absorption Values
  Vapor density vs air
  Transition temperature
  Potency
  Explosion limit
  Melting Point
  Restriction
  Refractive Index
  Vapor pressure
  Transmittance
  Quality clarifier
  Purity Grade
  Suitability
  Special Grade
Show More