• USA Home
  • Search Results

  • ethanolamine610814143511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ethanolamine

12 Products
Synonym:   2-Aminoethanol, 2-Aminoethyl alcohol, ETA, MEA 90, MEA-LCI, MEA, Monoethanolamine
Image of Ethanolamine
CAS Number:
Linear Formula:
 NH2CH2CH2OH
Molecular Weight:
 61.08
 Beilstein/REAXYS Number:
 505944
 EC Number:
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Restriction
  Refractive Index
  pK-Values
  Solubility
  Purification method
  Package quantity
  Biological Source
  Application(s)
  Viscosity
  Vapor pressure
  Transition temperature
  Melting Point
  Explosion limit
  Density
  Autoignition temp
  Physical Form
  pH Value
  Agency/Method
  Special Grade
  Product Line
  Potency
Show More