• USA Home
  • Search Results

  • ethanold65211151608111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ethanol-d6

2 Products
Synonym:   Ethyl alcohol-d6, Ethyl-d5 alcohol-d, Hexadeuteroethanol
Image of Ethanol-d<SUB>6</SUB>
CAS Number:
Linear Formula:
 CD3CD2OD
Molecular Weight:
 52.11
 Beilstein/REAXYS Number:
 1699096
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Explosion limit
  Assay
  Isotopic Purity
  Melting Point
Show More