• USA Home
  • Search Results

  • ethopabate237255906311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ethopabate

3 Products
Synonym:   Ethopabate, Ethyl pabate, Methyl 4-acetamido-2-ethoxybenzoate
Image of Ethopabate
CAS Number:
Empirical Formula (Hill Notation):
 C12H15NO4
Molecular Weight:
 237.25
 Beilstein/REAXYS Number:
 2808914
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Titertest
Show More