• USA Home
  • Search Results

  • ethopropazinehydrochloride34893109408211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ethopropazine hydrochloride

1 Products
Synonym:   10-(2-Diethylaminopropyl)phenothiazine, 10-[2-(Diethylamino)propyl]phenothiazine hydrochloride, Dibutil hydrochloride, Parkin
Image of Ethopropazine hydrochloride
CAS Number:
Empirical Formula (Hill Notation):
 C19H24N2S · HCl
Molecular Weight:
 348.93
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More