• USA Home
  • Search Results

  • ethoxyacetylenesolution700992780011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ethoxyacetylene solution

1 Products
Image of Ethoxyacetylene solution
CAS Number:
Linear Formula:
 C2H5OC≡CH
Molecular Weight:
 70.09
 Beilstein/REAXYS Number:
 741882
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More