• USA Home
  • Search Results

  • ethyl2bromocaprylate25116544529411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ethyl 2-bromocaprylate

1 Products
Synonym:   Ethyl 2-bromooctanoate
Image of Ethyl 2-bromocaprylate
CAS Number:
Linear Formula:
 CH3(CH2)5CHBrCOOC2H5
Molecular Weight:
 251.16
 Beilstein/REAXYS Number:
 1767535
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More