• USA Home
  • Search Results

  • ethyl2hydroxyhexanoate160215208955111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ethyl 2-hydroxyhexanoate

1 Products
Synonym:   Ethyl 2-hydroxycaproate
Image of Ethyl 2-hydroxyhexanoate
CAS Number:
Linear Formula:
 CH3(CH2)3CH(OH)COOC2H5
Molecular Weight:
 160.21
 Beilstein/REAXYS Number:
 1704446
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More