• USA Home
  • Search Results

  • ethyl2methyl4pentenoate142205339981811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ethyl 2-methyl-4-pentenoate

2 Products
Image of Ethyl 2-<WBR>methyl-<WBR>4-<WBR>pentenoate
CAS Number:
Linear Formula:
 H2C=CHCH2CH(CH3)CO2C2H5
Molecular Weight:
 142.20
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Assay
Show More