• USA Home
  • Search Results

  • ethyl2oxo4phenylbutyrate206246492029211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ethyl 2-oxo-4-phenylbutyrate

1 Products
Image of Ethyl 2-<WBR>oxo-<WBR>4-<WBR>phenylbutyrate
CAS Number:
Linear Formula:
 C6H5CH2CH2COCOOCH2CH3
Molecular Weight:
 206.24
 Beilstein/REAXYS Number:
 2725083
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More