• USA Home
  • Search Results

  • ethyl2trans4cisdecadienoate19629302530711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ethyl 2-trans-4-cis-decadienoate

2 Products
Image of Ethyl 2-<I>trans</I>-4-<I>cis</I>-decadienoate
CAS Number:
Linear Formula:
 CH3(CH2)4CH=CHCH=CHCO2C2H5
Molecular Weight:
 196.29
 Beilstein/REAXYS Number:
 1724176
 EC Number:
 FEMA Number:
 3148
 Flavis number:
 9.260
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Refractive Index
  Additive
  Application(s)
  Biological Source
  Agency/Method
  Document Links
  Allergen
  Organoleptic property
  Assay
Show More