• USA Home
  • Search Results

  • ethyl3aminobenzoatemethanesulfonatesalt2612988686211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ethyl 3-aminobenzoate methanesulfonate salt

1 Products
Synonym:   MS-222, Tricaine
Image of Ethyl 3-aminobenzoate methanesulfonate salt
CAS Number:
Empirical Formula (Hill Notation):
 C9H11NO2 · CH4SO3
Molecular Weight:
 261.29
 Beilstein/REAXYS Number:
 3775022
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID:
Popular Documents:  Product Information Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
Show More