• USA Home
  • Search Results

  • ethyl3ethoxyacrylate14417100126911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ethyl 3-ethoxyacrylate

1 Products
Synonym:   Ethyl 3-ethoxy-2-propenoate
Image of Ethyl 3-ethoxyacrylate
CAS Number:
Linear Formula:
 CH3CH2OCH=CHCOOCH2CH3
Molecular Weight:
 144.17
 Beilstein/REAXYS Number:
 1721612
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More