• USA Home
  • Search Results

  • ethyl3hydroxybutyrate13216540541411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ethyl 3-hydroxybutyrate

4 Products
Synonym:   grape butyrate
Image of Ethyl 3-hydroxybutyrate
CAS Number:
Linear Formula:
 CH3CH(OH)CH2COOC2H5
Molecular Weight:
 132.16
 Beilstein/REAXYS Number:
 1446190
 Council of Europe no.:
 10596
 EC Number:
 FEMA Number:
 3428
 Flavis number:
 9.522
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Document Links
  Allergen
  Organoleptic property
  Application(s)
  Assay
  Agency/Method
Show More