• USA Home
  • Search Results

  • ethyl3phenylpropionate17823202128511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ethyl 3-phenylpropionate

1 Products
Synonym:   Ethyl 3-phenylpropionate, Ethyl hydrocinnamate
Image of Ethyl 3-phenylpropionate
CAS Number:
Linear Formula:
 C6H5CH2CH2COOC2H5
Molecular Weight:
 178.23
 Beilstein/REAXYS Number:
 1910432
 Council of Europe no.:
 429
 EC Number:
 FEMA Number:
 2455
 Flavis number:
 9.747
 MDL number:
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
Show More