• USA Home
  • Search Results

  • ethyl4dimethylaminobenzoate193241028753311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ethyl 4-(dimethylamino)benzoate

3 Products
Synonym:   Parbenate
Image of Ethyl 4-<WBR>(dimethylamino)<WBR>benzoate
CAS Number:
Linear Formula:
 (CH3)2NC6H4CO2C2H5
Molecular Weight:
 193.24
 Beilstein/REAXYS Number:
 2210233
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Special Grade
Show More