• USA Home
  • Search Results

  • ethylacetate881114178611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ethyl acetate

32 Products
Synonym:   EtOAc
Image of Ethyl acetate
CAS Number:
Linear Formula:
 CH3COOC2H5
Molecular Weight:
 88.11
 Beilstein/REAXYS Number:
 506104
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Document Links
  Anal. Standard Form
  Shelf Life
  Allergen
  Organoleptic property
  Quality clarifier
  Package quantity
  Ice Shipping
  Density
  Restriction
  UV-Absorption Condition
  Color
  Purity Grade
  Product Line
  Refractive Index
  Suitability
  Transition temperature
  Agency/Method
  Potency
  Application(s)
  UV-Absorption Values
  Solubility
  Physical Form
  Special Grade
Show More

Product #
Description
270989 Ethyl acetate, anhydrous, 99.8%,
319902 Ethyl acetate, ACS reagent, ≥99.5%,
34858 Ethyl acetate, suitable for HPLC, ≥99.7%,
33211 Ethyl acetate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC),
650528 Ethyl acetate, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%,
27227 Ethyl acetate, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99.5% (GC),
58958 Ethyl acetate, analytical standard,
8.22277 Ethyl acetate, EMPLURA®,
EX0240 Ethyl acetate, Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS,
439169 Ethyl acetate, suitable for HPLC, ≥99.8%,
1.09623 Ethyl acetate, for analysis EMSURE® ACS,ISO,Reag. Ph Eur,
1.00789 Ethyl acetate, for gas chromatography MS SupraSolv®,
W241407 Ethyl acetate, ≥99%, FCC, FG,
1.10972 Ethyl acetate, for gas chromatography ECD and FID SupraSolv®,
1.03649 Ethyl acetate, hypergrade for LC-MS LiChrosolv®,
EX0245 Ethyl acetate, HPLC, Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs,
EX0241 Ethyl acetate, OmniSolv®,
1.00868 Ethyl acetate, for liquid chromatography LiChrosolv®,
537446 Ethyl acetate, ReagentPlus®, ≥99.8%,
W241415 Ethyl acetate, natural, ≥99%, FCC, FG,
1.00864 Ethyl acetate, EMPROVE® ESSENTIAL Ph Eur,BP,NF,
437549 Ethyl acetate, ACS reagent, ≥99.5%,
1265402 Ethyl acetate, United States Pharmacopeia (USP) Reference Standard,
1.00863 Ethyl acetate, for spectroscopy Uvasol®,
676810 Ethyl acetate, ACS reagent, ≥99.5%,
494518 Ethyl acetate, biotech. grade, ≥99.8%,
45767 Ethyl acetate, puriss. p.a., free of higher boiling impurities, ≥99.9% (GC),
270520 Ethyl acetate, HPLC grade, 99.8%,
L092004 Ethyl acetate,
L092003 Ethyl acetate,
PHR1481 Ethyl Acetate, Pharmaceutical Secondary Standard; Certified Reference Material,
RTC000087 Ethyl Acetate, Pharmaceutical Secondary Standard; Certified Reference Material,