• USA Home
  • Search Results

  • ethylacetimidatehydrochloride12358220807311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ethyl acetimidate hydrochloride

1 Products
Image of Ethyl acetimidate hydrochloride
CAS Number:
Linear Formula:
 CH3C(=NH)OC2H5 · HCl
Molecular Weight:
 123.58
 Beilstein Registry Number:
 3552401
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More