• USA Home
  • Search Results

  • ethylacrylate1001214088511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ethyl acrylate

4 Products
Synonym:   Acrylic acid ethyl ester
Image of Ethyl acrylate
CAS Number:
Linear Formula:
 CH2=CHCOOC2H5
Molecular Weight:
 100.12
 Beilstein/REAXYS Number:
 773866
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Refractive Index
  Document Links
  Allergen
  Organoleptic property
  Titertest
  Potency
  Biological Source
  Density
  Special Grade
  Additive
Show More