• USA Home
  • Search Results

  • ethylalcoholpure46076417511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ethyl alcohol, Pure

10 Products
Synonym:   Ethanol, Tax-free ethanol, absolute alcohol, non-denatured ethanol
Image of Ethyl alcohol, Pure
CAS Number:
Linear Formula:
 CH3CH2OH
Molecular Weight:
 46.07
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  UV-Absorption Values
  Suitability
  UV-Absorption Condition
  Solubility
  Color
  Density
  Restriction
  Special Grade
  Assay
  Vapor density vs air
  Melting Point
  Explosion limit
  Concentration
Show More

Product #
Description
E7023 Ethyl alcohol, Pure, 200 proof, for molecular biology,
459844 Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%,
459836 Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%,
E7148 Ethyl alcohol, Pure, 190 proof, for molecular biology,
493546 Ethyl alcohol, Pure, 200 proof, meets USP testing specifications,
459828 Ethyl alcohol, Pure, 200 proof, HPLC/spectrophotometric grade,
493511 Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%,
493538 Ethyl alcohol, Pure, 190 proof, meets USP testing specifications,
792780 Ethyl alcohol, Pure, 200 proof, ACS reagent, meets USP testing specifications, Excise Tax-free, Permit for use required,
792799 Ethyl alcohol, Pure, 190 proof, ACS reagent, meets USP testing specifications, Excise Tax-free, Permit for use required,