• USA Home
  • Search Results

  • ethylaminesolution45087504711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ethylamine solution

3 Products
Synonym:   Aminoethane, Monoethylamine
Image of Ethylamine solution
CAS Number:
Linear Formula:
 C2H5NH2
Molecular Weight:
 45.08
 Beilstein/REAXYS Number:
 505933
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
  Refractive Index
  Density
  Vapor pressure
Show More