• USA Home
  • Search Results

  • ethylazidoacetatesolution1291263781011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ethyl azidoacetate solution

3 Products
Image of Ethyl azidoacetate solution
CAS Number:
Linear Formula:
 N3CH2COOCH2CH3
Molecular Weight:
 129.12
 Beilstein/REAXYS Number:
 4247209
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
  Reaction Suitability
Show More