• USA Home
  • Search Results

  • ethylbcarboline3carboxylate240267421462311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ethyl β-carboline-3-carboxylate

1 Products
Synonym:   β-CCE, Ethyl 9H-pyrido[3,4-b]indole-3-carboxylate
Image of Ethyl β-carboline-3-carboxylate
CAS Number:
Empirical Formula (Hill Notation):
 C14H12N2O2
Molecular Weight:
 240.26
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More