• USA Home
  • Search Results

  • ethylbenzene1061710041411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ethylbenzene

8 Products
Image of Ethylbenzene
CAS Number:
Empirical Formula (Hill Notation):
 C8H10
Molecular Weight:
 106.17
 EC Index Number
 601-023-00-4
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Assay
Show More