• USA Home
  • Search Results

  • ethylbenzenesolution1061710041411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ethylbenzene solution

22 Products
Image of Ethylbenzene solution
CAS Number:
Empirical Formula (Hill Notation):
 C8H10
Molecular Weight:
 106.17
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Application(s)
  Anal. Standard Form
  Chemical Composition
  Concentration
  Composite Dimension
Show More

Product #
Description
487104 Ethylbenzene solution, NMR reference standard, 0.1% in chloroform-d (99.8 atom % D), TMS 0.01 %, NMR tube size 5 mm × 8 in.,
714917 Ethylbenzene solution, NMR reference standard, 0.1% in chloroform-d (99.8 atom % D), TMS 0.01 %, NMR tube size 5 mm × 8 in. , ultra-thin wall,
708046 Ethylbenzene solution, NMR reference standard, 10% in chloroform-d (99.8 atom % D),
736198 Ethylbenzene solution, NMR reference standard, 10% in chloroform-d (99.8 atom % D), NMR tube size 5 mm × 8 in. , ultra-thin wall,
40036 Ethylbenzene solution, certified reference material, 5000 μg/mL in methanol,
717959 Ethylbenzene solution, NMR reference standard, 1% in chloroform-d (99.8 atom % D),
487112 Ethylbenzene solution, NMR reference standard, 0.1% in chloroform-d (99.8 atom % D), TMS 0.01 %, NMR tube size 3 mm × 8 in.,
717967 Ethylbenzene solution, NMR reference standard, 0.1% in chloroform-d (99.8 atom % D), TMS 0.1 %,
612529 Ethylbenzene solution, NMR reference standard, 10% in chloroform-d (99.8 atom % D), TMS 0.01 %, NMR tube size 5 mm × 8 in.,
612758 Ethylbenzene solution, NMR reference standard, 0.4% in chloroform-d (99.8 atom %), NMR tube size 3 mm × 8 in.,
612456 Ethylbenzene solution, NMR reference standard, 0.1% in chloroform-d (99.8 atom % D), NMR tube size 8 mm × 7 in.,
733768 Ethylbenzene solution, NMR reference standard, 10% in chloroform-d (99.8 atom % D), NMR tube size 10 mm × 8 in.,
721018 Ethylbenzene solution, NMR reference standard, 0.1% in chloroform-d (99.8 atom % D), NMR tube size 5 mm × 8 in.,
721026 Ethylbenzene solution, NMR reference standard, 0.1% in chloroform-d (99.8 atom % D), NMR tube size 3 mm × 8 in.,
720291 Ethylbenzene solution, NMR reference standard, 10% in chloroform-d (99.8 atom % D), NMR tube size 3 mm × 8 in.,
710725 Ethylbenzene solution, NMR reference standard, 5% in chloroform-d (99.8 atom % D), TMS 1 %, NMR tube size 5 mm × 7 in.,
708054 Ethylbenzene solution, NMR reference standard, 0.1% in chloroform-d ("100%", 99.96 atom % D),
612936 Ethylbenzene solution, NMR reference standard, 5% in chloroform-d (99.8 atom % D), TMS 2 %, NMR tube size 3 mm × 8 in.,
612065 Ethylbenzene solution, NMR reference standard, 0.1% in chloroform-d (99.8 atom % D), TMS 0.01 %, NMR tube size 10 mm × 8 in.,
612480 Ethylbenzene solution, NMR reference standard, 0.1% in chloroform-d (99.8 atom % D), chromium(III) acetylacetonate 0.1 mg/mL, TMS 0.01 %, NMR tube size 5 mm × 8 in.,
487120 Ethylbenzene solution, NMR reference standard, 0.1% in chloroform-d (99.8 atom % D), TMS 0.01 %, NMR tube size 8 mm × 8 in.,
551341 Ethylbenzene solution, NMR reference standard, 5% in chloroform-d (99.8 atom % D), TMS 2 %, NMR tube size 5 mm × 8 in.,