• USA Home
  • Search Results

  • ethylbenzoylacetate192219402011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ethyl benzoylacetate

6 Products
Synonym:   Benzoylacetic acid ethyl ester, 3-Oxo-3-phenylpropionic acid ethyl ester, Ethyl benzoylacetate
Image of Ethyl benzoylacetate
CAS Number:
Empirical Formula (Hill Notation):
 C11H12O3
Molecular Weight:
 192.21
 EC Index Number:
 202-295-3
 MDL number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
  Refractive Index
  Manufacturer designation
  Special Grade
  Assay
Show More