• USA Home
  • Search Results

  • ethylcellulose12345900457311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ethyl cellulose

10 Products
Image of Ethyl cellulose
CAS Number:
 MDL number:
 NACRES
 NA.25
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
  Anal. Standard Form
  Manufacturer designation
  Special Grade
  Transition temperature
  Extent of labeling
  Assay
  Solubility
  Viscosity
Show More

Product #
Description
46070 Ethyl cellulose, 48.0-49.5% (w/w) ethoxyl basis,
247499 Ethyl cellulose, viscosity 100 cP, 5 % in toluene/ethanol 80:20(lit.), extent of labeling: 48% ethoxyl,
46080 Ethyl cellulose, 48.0-49.5% (w/w) ethoxyl basis,
200646 Ethyl cellulose, viscosity 4 cP, 5 % in toluene/ethanol 80:20(lit.), extent of labeling: 48% ethoxyl,
433837 Ethyl cellulose, viscosity 46 cP, 5 % in toluene/ethanol 80:20(lit.), extent of labeling: 48% ethoxyl,
200689 Ethyl cellulose, viscosity 10 cP, 5 % in toluene/ethanol 80:20(lit.), extent of labeling: 48% ethoxyl,
200697 Ethyl cellulose, viscosity 22 cP, 5 % in toluene/ethanol 80:20(lit.), extent of labeling: 48% ethoxyl,
200654 Ethyl cellulose, viscosity 300 cP, 5 % in toluene/ethanol 80:20(lit.), extent of labeling: 48% ethoxyl,
1265504 Ethyl cellulose, United States Pharmacopeia (USP) Reference Standard,
Y0001633 Ethyl cellulose, European Pharmacopoeia (EP) Reference Standard,