• USA Home
  • Search Results

  • ethylcyclopropanecarboxylate11414460607911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ethyl cyclopropanecarboxylate

1 Products
Image of Ethyl cyclopropanecarboxylate
CAS Number:
Linear Formula:
 C3H5CO2C2H5
Molecular Weight:
 114.14
 Beilstein Registry Number:
 2039752
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More