• USA Home
  • Search Results

  • ethylene28057485111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ethylene

3 Products
Synonym:   Ethene
Image of Ethylene
CAS Number:
Linear Formula:
 CH2=CH2
Molecular Weight:
 28.05
 Beilstein/REAXYS Number:
 1730731
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Purity Grade
  ID clarifier
Show More