• USA Home
  • Search Results

  • ethylenecarbonate88069649111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ethylene carbonate

5 Products
Synonym:   1,3-Dioxolan-2-one
Image of Ethylene carbonate
CAS Number:
Empirical Formula (Hill Notation):
 C3H4O3
Molecular Weight:
 88.06
 Beilstein/REAXYS Number:
 106249
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Special Grade
  Application(s)
  Potency
  Melting Point
  Explosion limit
Show More