• USA Home
  • Search Results

  • ethylenecarbonatesolution123459649111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ethylene carbonate solution

2 Products
Synonym:   1,3-Dioxolan-2-one, Glycol carbonate
Image of Ethylene carbonate solution
CAS Number:
Linear Formula:
 C3H4O3
 NACRES:
 NA.22
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
  Physical Form
  Density
Show More