• USA Home
  • Search Results

  • ethylenediamine601010715311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ethylenediamine

8 Products
Synonym:   1,2-Diaminoethane
Image of Ethylenediamine
CAS Number:
Linear Formula:
 NH2CH2CH2NH2
Molecular Weight:
 60.10
 Beilstein/REAXYS Number:
 605263
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
 eCl@ss:
 39030201
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Refractive Index
  Transition temperature
  Application(s)
  Purity Grade
  Special Grade
  Product Line
  Potency
  pH Value
  Physical Form
  Solubility
Show More