• USA Home
  • Search Results

  • ethylenediaminepalladiumiichloride237421502099211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(Ethylenediamine)palladium(II) chloride

2 Products
Synonym:   Dichloro(1,2-diaminoethane)palladium, Dichloro(ethylenediamine)palladium(II), NSC 153155, NSC 209490, Palladium(II) chloride ethylenediamine complex
Image of (Ethylenediamine)<WBR>palladium(II) chloride
CAS Number:
Linear Formula:
 Pd(H2NCH2CH2NH2)Cl2
Molecular Weight:
 237.42
 Beilstein/REAXYS Number:
 4934132
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More