• USA Home
  • Search Results

  • ethylenediaminesolution601010715311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ethylenediamine solution

1 Products
Synonym:   1,2-Diaminoethane
Image of Ethylenediamine solution
CAS Number:
Linear Formula:
 NH2CH2CH2NH2
Molecular Weight:
 60.10
 Beilstein/REAXYS Number:
 4123523
 MDL number:
 PubChem Substance ID:
 eCl@ss
 39030201
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More