• USA Home
  • Search Results

  • ethylenediaminetetraaceticacid292246000411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ethylenediaminetetraacetic acid

9 Products
Synonym:   (Ethylenedinitrilo)tetraacetic acid, EDTA, Edathamil, Ethylenedinitrilotetraacetic acid
Image of Ethylenediaminetetraacetic acid
CAS Number:
Linear Formula:
 (HO2CCH2)2NCH2CH2N(CH2CO2H)2
Molecular Weight:
 292.24
 Beilstein Registry Number:
 1716295
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  UV-Absorption Values
  Quality clarifier
  Product Line
  Solubility
  Physical Form
  Special Grade
Show More