• USA Home
  • Search Results

  • ethylenediaminetetraaceticaciddipotassiumsaltdihydrate404452510212911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ethylenediaminetetraacetic acid dipotassium salt dihydrate

4 Products
Synonym:   (Ethylenedinitrilo)tetraacetic acid dipotassium salt, Dipotassium ethylenediaminetetraacetate dihydrate, EDTA dipotassium salt, Edathamil, Potassium ethylenediaminetetraacetate dibasic
Image of Ethylenediaminetetraacetic acid dipotassium salt dihydrate
CAS Number:
Linear Formula:
 HOOCCH2(KOOCCH2)NCH2CH2N(CH2COOK)CH2COOH · 2H2O
Molecular Weight:
 404.45
 EC Number:
 NACRES
 NA.77
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Physical Form
  UV-Absorption Values
  Product Line
  pH Value
  Solubility
Show More