• USA Home
  • Search Results

  • ethylenediaminetetraaceticaciddisodiumsaltdihydrate37224638192611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ethylenediaminetetraacetic acid disodium salt dihydrate

10 Products
Synonym:   Disodium ethylenediaminetetraacetate dihydrate, EDTA disodium salt, EDTA-Na2, Edathamil, Edetate disodium salt dihydrate, Sequestrene Na2
Image of Ethylenediaminetetraacetic acid disodium salt dihydrate
CAS Number:
Empirical Formula (Hill Notation):
 C10H14N2Na2O8 · 2H2O
Molecular Weight:
 372.24
 Beilstein/REAXYS Number:
 3900609
 EC Number:
 NACRES:
 NA.31
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Quality clarifier
  Product Line
  Biological Source
  Restriction
  Agency/Method
  Application(s)
  UV-Absorption Values
  pH Value
  Physical Form
  Solubility
  Assay
Show More

Product #
Description
E5134 Ethylenediaminetetraacetic acid disodium salt dihydrate, for electrophoresis, for molecular biology, 99.0-101.0% (titration),
E4884 Ethylenediaminetetraacetic acid disodium salt dihydrate, ACS reagent, 99.0-101.0%,
ED2SS Ethylenediaminetetraacetic acid disodium salt dihydrate, reagent grade, 98.5-101.5% (titration),
E6635 Ethylenediaminetetraacetic acid disodium salt dihydrate, Sigma Grade, suitable for plant cell culture, 98.5-101.5%,
E1644 Ethylenediaminetetraacetic acid disodium salt dihydrate, BioUltra, 98.5-101.5%,
27285 Ethylenediaminetetraacetic acid disodium salt dihydrate, meets analytical specification of Ph. Eur., BP, USP, FCC, 99.0-101.0%,
03677 Ethylenediaminetetraacetic acid disodium salt dihydrate, BioUltra, for molecular biology, ≥99.0% (T),
03685 Ethylenediaminetetraacetic acid disodium salt dihydrate, ≥97.0% (KT),
PHR1068 Ethylenediaminetetraacetic acid disodium salt dihydrate, Pharmaceutical Secondary Standard; Certified Reference Material,
79884 Ethylenediaminetetraacetic acid disodium salt dihydrate, for HPLC, LiChropur, 99.0-101.0% (KT),