• USA Home
  • Search Results

  • ethylenediaminetetraaceticaciddisodiumsaltsolution3362113933311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ethylenediaminetetraacetic acid disodium salt solution

2 Products
Synonym:   (Ethylenedinitrilo)tetraacetic acid disodium salt, EDTA disodium salt, EDTA-Na2
Image of Ethylenediaminetetraacetic acid disodium salt solution
CAS Number:
Linear Formula:
 [-CH2N(CH2CO2Na)CH2CO2H]2
Molecular Weight:
 336.21
 Beilstein/REAXYS Number:
 3822669
 MDL number:
 NACRES:
 NA.31
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
  Restriction
Show More