• USA Home
  • Search Results

  • ethylenediaminetetraaceticacidsolution292246000411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ethylenediaminetetraacetic acid solution

2 Products
Synonym:   EDTA solution
Image of Ethylenediaminetetraacetic acid solution
CAS Number:
Empirical Formula (Hill Notation):
 C10H16N2O8
Molecular Weight:
 292.24
 Beilstein/REAXYS Number:
 1716295
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
 eCl@ss
 39030907
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
  Special Grade
  pH Value
  Product Line
Show More