• USA Home
  • Search Results

  • ethyleneglycol5msolution620710721111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ethylene glycol 5 M solution

1 Products
Synonym:   Additive Screening Solution 01/Kit-No 78374, Ethylene glycol solution
CAS Number:
Empirical Formula (Hill Notation):
 C2H6O2
Molecular Weight:
 62.07
 MDL number:
 NACRES:
 NA.32
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More