• USA Home
  • Search Results

  • ethyleneglycol620710721111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ethylene glycol

13 Products
Synonym:   1,2-Ethanediol
Image of Ethylene glycol
CAS Number:
Linear Formula:
 HOCH2CH2OH
Molecular Weight:
 62.07
 Beilstein/REAXYS Number:
 505945
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
 eCl@ss:
 39020501
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Restriction
  Refractive Index
  UV-Absorption Condition
  Application(s)
  Ice Shipping
  Explosion limit
  Density
  Autoignition temp
  Anal. Standard Form
  pH Value
  Vapor pressure
  Transition temperature
  Solubility
  Melting Point
  Physical Form
  UV-Absorption Values
  Assay
Show More