• USA Home
  • Search Results

  • ethyleneglycoldimethacrylate198229790511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ethylene glycol dimethacrylate

2 Products
Synonym:   1,2-Ethanediol dimethacrylate, Ethylene dimethacrylate
Image of Ethylene glycol dimethacrylate
CAS Number:
Linear Formula:
 CH2=C(CH3)COOCH2CH2OCOC(CH3)=CH2
Molecular Weight:
 198.22
 Beilstein/REAXYS Number:
 1776663
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Functional group
Show More