• USA Home
  • Search Results

  • ethyleneglycolmonomethylether760910986411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ethylene glycol monomethyl ether

2 Products
Synonym:   2-Methoxyethanol, Ethylene glycol monomethyl ether, Methyl glycol
Image of Ethylene glycol monomethyl ether
CAS Number:
Linear Formula:
 CH3OCH2CH2OH
Molecular Weight:
 76.09
 Beilstein/REAXYS Number:
 1731074
 EC Index Number:
 203-713-7
 MDL number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Physical Form
  pH Value
  Viscosity
  Transition temperature
Show More