• USA Home
  • Search Results

  • ethyleneglycolmonophenylether1381612299611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ethylene glycol monophenyl ether

2 Products
Synonym:   2-Phenoxyethanol, DOWANOL® EPh, Phenylglycol
Image of Ethylene glycol monophenyl ether
CAS Number:
Linear Formula:
 C6H5OCH2CH2OH
Molecular Weight:
 138.16
 Beilstein/REAXYS Number:
 1364011
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Potency
  Physical Form
  pH Value
Show More