• USA Home
  • Search Results

  • ethyleneoxidesolution44057521811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ethylene oxide solution

4 Products
Image of Ethylene oxide solution
CAS Number:
Empirical Formula (Hill Notation):
 C2H4O
Molecular Weight:
 44.05
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Anal. Standard Form
  Special Grade
  Purity Grade
  Package quantity
  Concentration
Show More