• USA Home
  • Search Results

  • ethylhexanoate1442112366011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ethyl hexanoate

4 Products
Synonym:   Caproic acid ethyl ester, Ethyl caproate
Image of Ethyl hexanoate
CAS Number:
Linear Formula:
 CH3(CH2)4COOC2H5
Molecular Weight:
 144.21
 Beilstein/REAXYS Number:
 1701293
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Document Links
  Special Grade
  Allergen
  Organoleptic property
  Agency/Method
  Assay
Show More