• USA Home
  • Search Results

  • ethyliodoacetate2140062348311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ethyl iodoacetate

1 Products
Image of Ethyl iodoacetate
CAS Number:
Linear Formula:
 ICH2COOC2H5
Molecular Weight:
 214.00
 Beilstein/REAXYS Number:
 741934
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More

Product #
Description
242934 Ethyl iodoacetate, 98%,