• USA Home
  • Search Results

  • ethyllinoleate3085054435411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ethyl linoleate

4 Products
Synonym:   Linoleic acid ethyl ester
Image of Ethyl linoleate
CAS Number:
Linear Formula:
 CH3(CH2)3(CH2CH=CH)2(CH2)7COOC2H5
Molecular Weight:
 308.50
 Beilstein/REAXYS Number:
 1727827
 EC Number:
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Titertest
Show More