• USA Home
  • Search Results

  • ethylllactate1181368747811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(−)-Ethyl L-lactate

4 Products
Synonym:   (−)-Ethyl (S)-2-hydroxypropionate, (S)-(−)-2-Hydroxypropionic acid ethyl ester, L-Lactic acid ethyl ester
Image of (−)-Ethyl <SC>L</SC>-lactate
CAS Number:
Empirical Formula (Hill Notation):
 C5H10O3
Molecular Weight:
 118.13
 Beilstein/REAXYS Number:
 1720839
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More